Learn More
Medchemexpress LLC Donitriptan 25mg | 170912-52-4 | 25MG
SDP
Catalog No. 5000244189
Encompass_Preferred
Donitriptan is a potent high efficacy agonist at 5-HT1B/1D receptors with pKis of 9 4 and 9 3 respectively[1
Catalog No. 50-002-44189 Supplier Medchemexpress LLC Supplier No. HY10615725MG
Donitriptan is a potent high efficacy agonist at 5-HT1B/1D receptors with pKis of 9 4 and 9 3 respectively[1