Learn More
Medchemexpress LLC Donitriptan (F-11356 free base) | 170912-52-4 | MFCD09832566 | 98.1% | 403.48 g/mol | C23H25N5O2 | 100 MG
SDP
Catalog No. 5000241856
Encompass_Preferred
Donitriptan is a research chemical that acts as a potent, high-efficacy agonist at 5-HT1B and 5-HT1D serotonin receptors. Supplied as the free base, it is intended for laboratory research use only and is not for human or veterinary use.
  • Potent 5-HT1B/1D receptor agonist with reported pKi 9.4 and 9.3
  • High purity (98.06%) suitable for biochemical and pharmacological assays
  • Solid form available in laboratory scales, for example 100 mg
  • Molecular weight 403.48 g/mol and formula C23H25N5O2 for reference
  • Datasheet, COA, and SDS available to support handling and characterization
Catalog No. 50-002-41856 Supplier Medchemexpress LLC Supplier No. HY106157100MG
Donitriptan is a research chemical that acts as a potent, high-efficacy agonist at 5-HT1B and 5-HT1D serotonin receptors. Supplied as the free base, it is intended for laboratory research use only and is not for human or veterinary use.
  • Potent 5-HT1B/1D receptor agonist with reported pKi 9.4 and 9.3
  • High purity (98.06%) suitable for biochemical and pharmacological assays
  • Solid form available in laboratory scales, for example 100 mg
  • Molecular weight 403.48 g/mol and formula C23H25N5O2 for reference
  • Datasheet, COA, and SDS available to support handling and characterization