Learn More
Immunoreagents Inc DONKEY A-GOAT IGG DYLIGHT 633
Catalog No. 50270907
Encompass_Preferred
Secondary Antibody; Donkey anti-goat IgG (H&L), Affinity Pure, DyLight 633 Conjugate, Host: Donkey, Format: Lyo, Label: DyL633, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-270-907 Supplier Immunoreagents Inc Supplier No. DKXGT003D633NHSX
Secondary Antibody; Donkey anti-goat IgG (H&L), Affinity Pure, DyLight 633 Conjugate, Host: Donkey, Format: Lyo, Label: DyL633, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS