Learn More
Immunoreagents Inc DONKEY A-GOAT IGG DYLIGHT 650
Catalog No. 50271975
Encompass_Preferred
Seconday Antibody; Donkey anti-goat IgG (H&L), Affinity Pure, min x w/Hu, Mu, Rb or Rt lgG/SP, DyLight 650 Conjugate, Host: Donkey, Format: Lyo, Label: DyL650, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-975 Supplier Immunoreagents Inc Supplier No. DKXGT003E650NHSX
Seconday Antibody; Donkey anti-goat IgG (H&L), Affinity Pure, min x w/Hu, Mu, Rb or Rt lgG/SP, DyLight 650 Conjugate, Host: Donkey, Format: Lyo, Label: DyL650, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS