Learn More
Immunoreagents Inc DONKEY A-GOAT IGG DYLIGHT 650
Catalog No. 50270908
Encompass_Preferred
Secondary Antibody; Donkey anti-goat IgG (H&L), Affinity Pure, DyLight 650 Conjugate, Host: Donkey, Format: Lyo, Label: DyL650, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-270-908 Supplier Immunoreagents Inc Supplier No. DKXGT003D650NHSX
Secondary Antibody; Donkey anti-goat IgG (H&L), Affinity Pure, DyLight 650 Conjugate, Host: Donkey, Format: Lyo, Label: DyL650, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS