Learn More
Immunoreagents Inc DONKEY A-GOAT IGG HRP 1 MG
Catalog No. 50271769
Encompass_Preferred
Seconday Antibody; Donkey anti-goat IgG (H&L), Affinity Pure, min x w/Hu, Mu, Rb or Rt lgG/SP, HRP Conjugate, Host: Donkey, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-769 Supplier Immunoreagents Inc Supplier No. DKXGT003EHRPX
Seconday Antibody; Donkey anti-goat IgG (H&L), Affinity Pure, min x w/Hu, Mu, Rb or Rt lgG/SP, HRP Conjugate, Host: Donkey, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC