Learn More
Immunoreagents Inc DONKEY A-GOAT IGG HRP 1MG
Catalog No. 50270903
Encompass_Preferred
Secondary Antibody; Donkey anti-goat IgG (H&L), Affinity Pure, HRP Conjugate, Host: Donkey, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-270-903 Supplier Immunoreagents Inc Supplier No. DKXGT003DHRPX
Secondary Antibody; Donkey anti-goat IgG (H&L), Affinity Pure, HRP Conjugate, Host: Donkey, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC