Learn More
Immunoreagents Inc DONKEY A-MOUSE IGG ALP
Catalog No. 50271789
Encompass_Preferred
Secondary Antibody; Donkey anti-mouse IgG (H&L) - F(ab)'2 fragment, min x w/Bo, Ck, Eq, Gt, Gp, Ha, Hu, Rb, Rt or Sh IgG, ALP Conjugate, Host: Donkey, Format: Liq, Label: ALP, Applications: WB, ELISA
Catalog No. 50-271-789 Supplier Immunoreagents Inc Supplier No. DKXMU003LALP
Secondary Antibody; Donkey anti-mouse IgG (H&L) - F(ab)'2 fragment, min x w/Bo, Ck, Eq, Gt, Gp, Ha, Hu, Rb, Rt or Sh IgG, ALP Conjugate, Host: Donkey, Format: Liq, Label: ALP, Applications: WB, ELISA