Learn More
Immunoreagents Inc DONKEY A-MOUSE IGG AP
Supplier:  Immunoreagents Inc DKXMU003L
Encompass_Preferred
Secondary Antibody; Donkey anti-mouse IgG (H&L) - F(ab)'2 fragment, min x w/Bo, Ck, Eq, Gt, Gp, Ha, Hu, Rb, Rt or Sh IgG, Host: Donkey, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-270-805