Learn More
Immunoreagents Inc DONKEY A-MOUSE IGG AP
Supplier: Immunoreagents Inc DKXMU003G
Secondary Antibody; Donkey anti-mouse IgG (H&L) - F(ab)'2 fragment, Host: Donkey, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC