Learn More
Immunoreagents Inc DONKEY A-MOUSE IGG DYLIGHT633
Catalog No. 50271929
Encompass_Preferred
Secondary Antibody; Donkey anti-mouse IgG (H&L) - Affinity Pure, min x w/Bo, Ck, Eq, Gt, GP, Ha, Hu, Rb, Rt or Sh IgG, DyLight 633 Conjugate, Host: Donkey, Format: Lyo, Label: DyL633, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-929 Supplier Immunoreagents Inc Supplier No. DKXMU003F633NHSX
Secondary Antibody; Donkey anti-mouse IgG (H&L) - Affinity Pure, min x w/Bo, Ck, Eq, Gt, GP, Ha, Hu, Rb, Rt or Sh IgG, DyLight 633 Conjugate, Host: Donkey, Format: Lyo, Label: DyL633, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS