Learn More
Immunoreagents Inc DONKEY A-MOUSE IGG DYLIGHT800
Catalog No. 50271937
Encompass_Preferred
Secondary Antibody; Donkey anti-mouse IgG (H&L) - F(ab)'2 fragment, DyLight 800 Conjugate, Host: Donkey, Format: Lyo, Label: DyL800, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-937 Supplier Immunoreagents Inc Supplier No. DKXMU003G800NHSX
Secondary Antibody; Donkey anti-mouse IgG (H&L) - F(ab)'2 fragment, DyLight 800 Conjugate, Host: Donkey, Format: Lyo, Label: DyL800, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS