Learn More
Immunoreagents Inc DONKEY A-MOUSE IGG FITC
Catalog No. 50271554
Encompass_Preferred
Secondary Antibody; Donkey anti-mouse IgG (H&L) - F(ab)'2 fragment, min x w/Bo, Ck, Eq, Gt, Gp, Ha, Hu, Rb, Rt or Sh IgG, FITC Conjugate, Host: Donkey, Format: Liq, Label: FITC, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-554 Supplier Immunoreagents Inc Supplier No. DKXMU003LFITC
Secondary Antibody; Donkey anti-mouse IgG (H&L) - F(ab)'2 fragment, min x w/Bo, Ck, Eq, Gt, Gp, Ha, Hu, Rb, Rt or Sh IgG, FITC Conjugate, Host: Donkey, Format: Liq, Label: FITC, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS