Learn More
Immunoreagents Inc DONKEY A-MOUSE IGG FITC
Supplier:  Immunoreagents Inc DKXMU003GFITC
Encompass_Preferred
Secondary Antibody; Donkey anti-mouse IgG (H&L) - F(ab)'2 fragment, FITC Conjugate, Host: Donkey, Format: Liq, Label: FITC, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-145