Learn More
Immunoreagents Inc DONKEY A-MOUSE IGG HRP
Supplier:  Immunoreagents Inc DKXMU003DHRPX
Encompass_Preferred
Secondary Antibody; Donkey anti-mouse IgG (H&L) - Affinity Pure, HRP Conjugate, Host: Donkey, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-272-120