Learn More
Immunoreagents Inc DONKEY A-RABBIT IGG ALP
Supplier:  Immunoreagents Inc DKXRB003FALP
Encompass_Preferred
Secondary Antibody; Donkey anti-rabbit IgG (H&L) - Affinity Pure, min x w/Bo, Ck, Eq, Gt, Gp, Ha, Hu, Mu, Rt or Sh IgG, ALP Conjugate, Host: Donkey, Format: Liq, Label: ALP, Applications: WB, ELISA
Catalog No. 50-271-943