Learn More
Immunoreagents Inc DONKEY A-RABBIT IGG BIO
Supplier:  Immunoreagents Inc DKXRB003EBIO
Encompass_Preferred
Secondary Antibody; Donkey anti-rabbit IgG (H&L) - Affinity Pure, min x w/Mouse IgG or Serum Proteins, Biotin Conjugate, Host: Donkey, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-750