Learn More
Immunoreagents Inc DONKEY A-RABBIT IGG BIO
Catalog No. 50271944
Encompass_Preferred
Secondary Antibody; Donkey anti-rabbit IgG (H&L) - Affinity Pure, min x w/Bo, Ck, Eq, Gt, Gp, Ha, Hu, Mu, Rt or Sh IgG, Biotin Conjugate, Host: Donkey, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-944 Supplier Immunoreagents Inc Supplier No. DKXRB003FBIO
Secondary Antibody; Donkey anti-rabbit IgG (H&L) - Affinity Pure, min x w/Bo, Ck, Eq, Gt, Gp, Ha, Hu, Mu, Rt or Sh IgG, Biotin Conjugate, Host: Donkey, Format: Liq, Label: Biotin, Applications: WB, ELISA, IHC/ICC