Learn More
Immunoreagents Inc DONKEY A-RABBIT IGG DYLIT350
Catalog No. 50272086
Encompass_Preferred
Secondary Antibody; Donkey anti-rabbit IgG (H&L) - Affinity Pure, min x w/Mouse IgG or Serum Proteins, DyLight 350 Conjugate, Host: Donkey, Format: Lyo, Label: DyL350, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-272-086 Supplier Immunoreagents Inc Supplier No. DKXRB003E350NHSX
Secondary Antibody; Donkey anti-rabbit IgG (H&L) - Affinity Pure, min x w/Mouse IgG or Serum Proteins, DyLight 350 Conjugate, Host: Donkey, Format: Lyo, Label: DyL350, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS