Learn More
Immunoreagents Inc DONKEY A-RABBIT IGG DYLIT550
Supplier:  Immunoreagents Inc DKXRB003E550NHSX
Encompass_Preferred
Secondary Antibody; Donkey anti-rabbit IgG (H&L) - Affinity Pure, min x w/Mouse IgG or Serum Proteins, DyLight 550 Conjugate, Host: Donkey, Format: Lyo, Label: DyL550, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-272-087