Learn More
Immunoreagents Inc DONKEY A-RABBIT IGG FITC
Catalog No. 50272020
Encompass_Preferred
Secondary Antibody; Donkey anti-rabbit IgG (H&L) - Affinity Pure, min x w/Mouse IgG or Serum Proteins, FITC Conjugate, Host: Donkey, Format: Liq, Label: FITC, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-272-020 Supplier Immunoreagents Inc Supplier No. DKXRB003EFITC
Secondary Antibody; Donkey anti-rabbit IgG (H&L) - Affinity Pure, min x w/Mouse IgG or Serum Proteins, FITC Conjugate, Host: Donkey, Format: Liq, Label: FITC, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS