Learn More
Immunoreagents Inc DONKEY A-RAT IGG ALP
Catalog No. 50271660
Encompass_Preferred
Secondary Antibody; Donkey anti-rat IgG (H&L) - Affinity Pure, ALP Conjugate, Host: Donkey, Format: Liq, Label: ALP, Applications: WB, ELISA
Catalog No. 50-271-660 Supplier Immunoreagents Inc Supplier No. DKXRT003DALP
Secondary Antibody; Donkey anti-rat IgG (H&L) - Affinity Pure, ALP Conjugate, Host: Donkey, Format: Liq, Label: ALP, Applications: WB, ELISA