Learn More
Immunoreagents Inc DONKEY A-RAT IGG DYLIGHT 594
Catalog No. 50271948
Encompass_Preferred
Secondary Antibody; Donkey anti-rat IgG (H&L) - Affinity Pure, min x w/Bo, Ck, Eq, Gt, Gp, Ha, Hu, Mu, Rb or Sh IgG, DyLight 594 Conjugate, Host: Donkey, Format: Lyo, Label: DyL594, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-948 Supplier Immunoreagents Inc Supplier No. DKXRT003F594NHSX
Secondary Antibody; Donkey anti-rat IgG (H&L) - Affinity Pure, min x w/Bo, Ck, Eq, Gt, Gp, Ha, Hu, Mu, Rb or Sh IgG, DyLight 594 Conjugate, Host: Donkey, Format: Lyo, Label: DyL594, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS