Learn More
Immunoreagents Inc DONKEY A-RAT IGG HRP
Catalog No. 50271353
Encompass_Preferred
Secondary Antibody; Donkey anti-rat IgG (H&L), F(ab)'2 fragment - Affinity Pure, min x w/Bo, Ck, Eq, Gt, Gp, Ha, Hu, Mu, Rb or Sh IgG, HRP Conjugate, Host: Donkey, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-271-353 Supplier Immunoreagents Inc Supplier No. DKXRT003LHRPX
Secondary Antibody; Donkey anti-rat IgG (H&L), F(ab)'2 fragment - Affinity Pure, min x w/Bo, Ck, Eq, Gt, Gp, Ha, Hu, Mu, Rb or Sh IgG, HRP Conjugate, Host: Donkey, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC