Learn More
Immunoreagents Inc DONKEY A-SHEEP IGG ALP
Supplier:  Immunoreagents Inc DKXSH003GALP
Encompass_Preferred
Secondary Antibody: Donkey anti-sheep IgG (H&L), F(ab)'2 fragment - Affinity Pure, ALP Conjugate, Host: Donkey, Format: Liq, Label: ALP, Applications: WB, ELISA
Catalog No. 50-271-355