Learn More
Immunoreagents Inc DONKEY A-SHEEP IGG AP
Supplier:  Immunoreagents Inc DKXSH003E
Encompass_Preferred
Secondary Antibody: Donkey anti-sheep IgG (H&L) - Affinity Pure, min x w/Hu or Rb IgG, Host: Donkey, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-271-024