Learn More
Immunoreagents Inc DONKEY A-SHEEP IGG AP
Supplier: Immunoreagents Inc DKXSH003F
Secondary Antibody: Donkey anti-sheep IgG (H&L) - Affinity Pure, min x w/Hu, Mu or Rb IgG, Host: Donkey, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC