Learn More
Immunoreagents Inc DONKEY A-SHEEP IGG DYLIGHT550
Catalog No. 50271816
Encompass_Preferred
Secondary Antibody: Donkey anti-sheep IgG (H&L) - Affinity Pure, min x w/Hu or Rb IgG, DyLight 550 Conjugate, Host: Donkey, Format: Lyo, Label: DyL550, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-816 Supplier Immunoreagents Inc Supplier No. DKXSH003E550NHSX
Secondary Antibody: Donkey anti-sheep IgG (H&L) - Affinity Pure, min x w/Hu or Rb IgG, DyLight 550 Conjugate, Host: Donkey, Format: Lyo, Label: DyL550, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS