Learn More
Immunoreagents Inc DONKEY A-SHEEP IGG DYLIGHT594
Catalog No. 50271963
Encompass_Preferred
Secondary Antibody: Donkey anti-sheep IgG (H&L) - Affinity Pure, min x w/Hu, Mu or Rb IgG, DyLight 594 Conjugate, Host: Donkey, Format: Lyo, Label: DyL594, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-963 Supplier Immunoreagents Inc Supplier No. DKXSH003F594NHSX
Secondary Antibody: Donkey anti-sheep IgG (H&L) - Affinity Pure, min x w/Hu, Mu or Rb IgG, DyLight 594 Conjugate, Host: Donkey, Format: Lyo, Label: DyL594, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS