Learn More
Immunoreagents Inc DONKEY A-SHEEP IGG DYLIGHT650
Catalog No. 50271819
Encompass_Preferred
Secondary Antibody: Donkey anti-sheep IgG (H&L) - Affinity Pure, min x w/Hu or Rb IgG, DyLight 650 Conjugate, Host: Donkey, Format: Lyo, Label: DyL650, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-271-819 Supplier Immunoreagents Inc Supplier No. DKXSH003E650NHSX
Secondary Antibody: Donkey anti-sheep IgG (H&L) - Affinity Pure, min x w/Hu or Rb IgG, DyLight 650 Conjugate, Host: Donkey, Format: Lyo, Label: DyL650, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS