Learn More
Immunoreagents Inc DONKEY A-SHEEP IGG HRP
Supplier:  Immunoreagents Inc DKXSH003FHRPX
Encompass_Preferred
Secondary Antibody: Donkey anti-sheep IgG (H&L) - Affinity Pure, min x w/Hu, Mu or Rb IgG, HRP Conjugate, Host: Donkey, Format: Lyo, Label: HRP, Applications: WB, ELISA, IHC/ICC
Catalog No. 50-272-036