Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc Donkey aGt IgG Anti Secondary

Supplier:  Cytek Biosciences Inc 728082M001

Encompass_Preferred

Product Name: HRP Donkey Anti-Goat IgG (H&L)
Product Class: Antibody
Type: Secondary
Product Name: HRP Donkey Anti-Goat IgG (H&L)
Cytek Catalog Number: 72-8082-M001
Marker: IgG (H&L)
Target Species: Goat
Reported Cross Reactivity: Goat
Clonality: Polyclonal
Host Species: Donkey
Conjugation: HRP
Size: 1 mg

Catalog No. 50-000-87338


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.