Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgY Donkey anti-Chicken, Cy3, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP194CMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP194CMI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP194CMI Supplier MilliporeSigma Supplier No. AP194C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Donkey Polyclonal Antibody

Detect Donkey Chicken IgY using this Donkey anti-Chicken IgY Antibody, Cy3 conjugate, Species Adsorbed validated for use in immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Donkey anti-Chicken IgY Species adsorbed
Applications Immunofluorescence
Classification Polyclonal
Conjugate Cyanine3
Host Species Donkey
Quantity 500 μg
Primary or Secondary Secondary
Target Species Chicken

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.