Learn More
Jackson Immuno Research Labs Alexa Fluor 647 AffiniPure F(ab')₂ Fragment Donkey Anti-Goat IgG (H+L), .3mg
SDP
Catalog No. NC0356506
Encompass_Preferred
Alexa Fluor 647-AffiniPure F(ab')2 Fragment Donkey Anti-Goat IgG (H+L) (min X Ck,GP,Sy Hms,Hrs,Hu,Ms,Rb,Rat Sr Prot)
Catalog No. NC0356506 Supplier Jackson Immuno Research Labs Supplier No. 705606147
Alexa Fluor 647-AffiniPure F(ab')2 Fragment Donkey Anti-Goat IgG (H+L) (min X Ck,GP,Sy Hms,Hrs,Hu,Ms,Rb,Rat Sr Prot)