Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Donkey Anti-Goat IgG (H&L) Antibody (FITC), ProSci
SDP

Catalog No. 16817897 Shop All ProSci Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-817-897 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-817-897 Supplier ProSci LLC Supplier No. XR93602000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Secondary Antibody

Donkey Anti-Goat IgG (H&L) Antibody specifically detects Goat Anti-Mouse IgG (H&L) in Goat Samples. It is validated for ELISA, Western Blot, Immunohistochemistry.

Specifications

Antigen Donkey Anti-Goat IgG (H&L)
Applications ELISA, Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate FITC
Dilution Fluorescein Conjugated Affinity Purified anti-Goat IgG is suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, 0.01% (w/v) Sodium Azide; pH 7.2; 10 mg/mL Bovine Serum Albumin
Host Species Donkey
Immunogen Goat IgG - whole molecule
Purification Method Fluorescein Conjugated Affinity Purified anti-Goat IgG was prepared from monospecific antiserum by immunoaffinity chromatography using Goat IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Fluorescein, anti-Donkey Serum, Goat IgG, and Goat Serum.
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Goat
Content And Storage Alkaline Phosphatase Conjugated Affinity Purified anti-Rabbit IgG can be stored vial at 4 °C before opening. DO NOT FREEZE. This product is stable at 4 °C as an undiluted liquid. Dilute only prior to immediate use. Freezing alkaline phosphatase conjugates will result in a substantial loss of enzymatic activity. Expiration date is one (1) year from date of opening product.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.