Learn More
Immunology Consultants anti-Goat IgG h+l Antibody (Donkey) - Affinity Purified
SDP
Catalog No. 502248871
Encompass_Preferred
This affinity‑purified donkey antibody provides broad recognition of goat IgG by targeting both heavy and light chains, enabling sensitive detection of a wide range of goat primary antibodies. Its strong specificity and low background make it suitable for ELISA, Western blotting, immunofluorescence, and other immunoassay formats. The donkey host species minimizes cross‑reactivity with common mammalian proteins, supporting clean, high‑signal detection even in complex matrices. DGHL‑50A offers reliable lot‑to‑lot consistency and robust assay performance, making it a valuable secondary reagent for laboratories using goat‑derived primaries across diverse applications.
This item is not returnable. View return policy
Catalog No. 50-224-8871 Supplier Immunology Consultants Supplier No. DGHL50A
This affinity‑purified donkey antibody provides broad recognition of goat IgG by targeting both heavy and light chains, enabling sensitive detection of a wide range of goat primary antibodies. Its strong specificity and low background make it suitable for ELISA, Western blotting, immunofluorescence, and other immunoassay formats. The donkey host species minimizes cross‑reactivity with common mammalian proteins, supporting clean, high‑signal detection even in complex matrices. DGHL‑50A offers reliable lot‑to‑lot consistency and robust assay performance, making it a valuable secondary reagent for laboratories using goat‑derived primaries across diverse applications.