Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Donkey anti-Goat, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP180PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP180PMI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP180PMI Supplier MilliporeSigma Supplier No. AP180P
Add to Cart
Add to Cart

Donkey Polyclonal Antibody

Donkey anti-Goat IgG Antibody, HRP conjugate, Species Adsorbed is an antibody against Donkey Goat IgG for use in ELISA, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Donkey anti-Goat IgG Species adsorbed
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate HRP
Formulation Purified donkey secondary antibody IgG in lyophilized buffer 0.01M Sodium Phosphate, 0.15M NaCl, pH 7.1 with 15mg/mL BSA. Reconstitution: Reconstitute with 500μL of sterile distilled water.
Host Species Donkey
Purification Method Immunoaffinity Purified and Immunodepletion
Quantity 500 μL
Primary or Secondary Secondary
Target Species Goat
Content And Storage 2-8°C for up to 12 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.