Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Donkey anti-Goat, Rhodamine (TRITC), Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP180RMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP180RMI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP180RMI Supplier MilliporeSigma Supplier No. AP180R
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Donkey Polyclonal Antibody

Donkey anti-Goat IgG Antibody, Rhodamine conjugate, Species Adsorbed is an antibody against Donkey Goat IgG for use in immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Donkey anti-Goat IgG Species adsorbed
Applications Immunofluorescence
Classification Polyclonal
Conjugate Rhodamine
Formulation Lyophilized. Buffer = 0.02M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA, and 0.1% sodium azide. RECONSTITUTION: Reconstitute with 500μL of sterile distilled water. RHODAMINE/PROTEIN: Approximately 0.5 (A550/A280).
Host Species Donkey
Quantity 500 μg
Primary or Secondary Secondary
Target Species Goat
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.