Learn More
Jackson Immuno Research Labs Alexa Fluor 647 AffiniPure Donkey Anti-Guinea Pig IgG (H+L), .5mg
SDP
Catalog No. NC0292731
Encompass_Preferred
Alexa Fluor 647-AffiniPure Donkey Anti-Guinea Pig IgG (H+L) (min X Bov,Ck,Gt,Sy Hms,Hrs,Hu,Ms,Rb,Rat,Shp Sr Prot) Physical State: Freeze-dried powder Purity: The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. Buffer: 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 Stabilizer: 15 mg/ml Bovine Serum Albumin (IgG-Free
Catalog No. NC0292731 Supplier Jackson Immuno Research Labs Supplier No. 706605148
Alexa Fluor 647-AffiniPure Donkey Anti-Guinea Pig IgG (H+L) (min X Bov,Ck,Gt,Sy Hms,Hrs,Hu,Ms,Rb,Rat,Shp Sr Prot) Physical State: Freeze-dried powder Purity: The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. Buffer: 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 Stabilizer: 15 mg/ml Bovine Serum Albumin (IgG-Free