Learn More
Jackson Immuno Research Labs Cy™5 AffiniPure™ Donkey Anti-Human IgG (H+L), 0.5 mg
SDP
Catalog No. NC2865129
Encompass_Preferred
Target: HumanHost: DonkeyAntibody Format: Whole IgGSpecificity: IgG (H+L)Minimal Cross Reactivity: Bovine, Chicken, Goat, Guinea Pig, Syrian Hamster, Horse, Mouse, Rabbit, Rat, Sheep Serum ProteinsConjugate: Cyanine Cy™5Product Category: Cyanine Conjugates for Permanent MountingClonality: PolyclonalRRID: AB_2340539
Catalog No. NC2865129 Supplier Jackson Immuno Research Labs Supplier No. 709175149
Target: HumanHost: DonkeyAntibody Format: Whole IgGSpecificity: IgG (H+L)Minimal Cross Reactivity: Bovine, Chicken, Goat, Guinea Pig, Syrian Hamster, Horse, Mouse, Rabbit, Rat, Sheep Serum ProteinsConjugate: Cyanine Cy™5Product Category: Cyanine Conjugates for Permanent MountingClonality: PolyclonalRRID: AB_2340539