Learn More
Jackson Immuno Research Labs DONKEY ANTI-HUMAN IGM, AP CONJ
SDP
Catalog No. NC1707796
Encompass_Preferred
Alkaline Phosphatase AffiniPure F(ab')₂ Fragment Donkey Anti-Human IgM, Fc5μ fragment specific.Conjugate: Alkaline PhosphataseProduct Category: F(ab')₂ Fragment Affinity-Purified AntibodiesClonality: Polyclonal.
Catalog No. NC1707796 Supplier Jackson Immuno Research Labs Supplier No. 709056073
Alkaline Phosphatase AffiniPure F(ab')₂ Fragment Donkey Anti-Human IgM, Fc5μ fragment specific.Conjugate: Alkaline PhosphataseProduct Category: F(ab')₂ Fragment Affinity-Purified AntibodiesClonality: Polyclonal.