Learn More
Jackson Immuno Research Labs Cy™3 AffiniPure Donkey Anti-Mouse IgG (H+L), .5mg
SDP
Supplier:  Jackson Immuno Research Labs 715165150
Encompass_Preferred
Cy3-AffiniPure Donkey Anti-Mouse IgG (H+L) (min X Bov,Ck,Gt,GP,Sy Hms,Hrs,Hu,Rb,Shp Sr Prot)
Catalog No. NC9606617