Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Donkey anti-Mouse, Alkaline Phosphatase, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP192AMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP192AMI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP192AMI Supplier MilliporeSigma Supplier No. AP192A
Add to Cart
Add to Cart

Donkey Polyclonal Antibody

This Donkey anti-Mouse IgG Antibody, Alkaline Phosphatase conjugate, Species Adsorbed is validated for use in ELISA, western blotting for the detection of Donkey Mouse IgG.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Donkey anti-Mouse IgG Species adsorbed
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.6mg/mL
Conjugate Alkaline Phosphatase (AP)
Formulation Lyophilized. Buffer = 0.01M Tris-HCl, 0.25M NaCl, pH 8.0, with 15mg/mL BSA and 0.05% Sodium Azide. RECONSTITUTION: Reconstitute to 1.0mg/mL with 0.5mLsterile distilled water.
Host Species Donkey
Quantity 500 μL
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.