Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Donkey anti-Mouse IgG (H+L) ReadyProbes™ Secondary Antibody, Alexa Fluor™ 488

Catalog No. R37114
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 2.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R37114 2 x 2.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R37114 Supplier Invitrogen™ Supplier No. R37114
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Donkey Polyclonal Secondary Antibody

No pipetting or dilutions required. Just tip and drip two drops per mL buffer to stain your cells. This antibody shows minimum cross-reactivity to bovine, chicken, goat, guinea pig, hamster, horse, human, rabbit, rat, and sheep serum proteins. Like other ReadyProbes reagents, Alexa Fluor 488 Donkey anti-Mouse IgG Antibody comes in a convenient dropper bottle.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG (H+L)
Applications Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry, Multiplex Immunohistochemistry
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation PBS with 0.1% BSA and <0.1% sodium azide
Host Species Donkey
Immunogen Gamma Immunoglobin.
Purification Method Affinity chromatography
Quantity 2 x 2.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 4°C, store in dark
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.