Learn More
Jackson Immuno Research Labs Cy™3 AffiniPure Donkey Anti-Rabbit IgG (H+L)(min X Bov, Ck, Gt, GP, Sy Hms, Hrs, Hu, Ms, Rat, Shp Sr Prot)
SDP
Catalog No. NC9805921
Encompass_Preferred
Whole IgG antibodies are isolated as intact molecules from antisera by immunoaffinity chromatography. They have an Fc portion and two antigen binding Fab portions joined together by disulfide bonds and therefore they are divalent. The average molecular weight is reported to be about 160 kDa. The whole IgG form of antibodies is suitable for the majority of immunodetection procedures and is the most cost effective. Purity: The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. Buffer: 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6
Catalog No. NC9805921 Supplier Jackson Immuno Research Labs Supplier No. 711165152
Whole IgG antibodies are isolated as intact molecules from antisera by immunoaffinity chromatography. They have an Fc portion and two antigen binding Fab portions joined together by disulfide bonds and therefore they are divalent. The average molecular weight is reported to be about 160 kDa. The whole IgG form of antibodies is suitable for the majority of immunodetection procedures and is the most cost effective. Purity: The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. Buffer: 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6