Learn More
Jackson Immuno Research Labs Rhodamine (TRITC) AffiniPure Donkey Anti-Rabbit IgG (H+L), .5mg
SDP
Catalog No. NC9937469
Encompass_Preferred
Rhodamine (TRITC)-AffiniPure Donkey Anti-Rabbit IgG (H+L) (min X Bov,Ck,Gt,GP,Sy Hms,Hrs,Hu,Ms,Rat,Shp Sr Prot)
Catalog No. NC9937469 Supplier Jackson Immuno Research Labs Supplier No. 711025152
Rhodamine (TRITC)-AffiniPure Donkey Anti-Rabbit IgG (H+L) (min X Bov,Ck,Gt,GP,Sy Hms,Hrs,Hu,Ms,Rat,Shp Sr Prot)