Learn More
Invitrogen™ Donkey anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 488
Shop All Thermo Scientific ProductsDescription
This antibody is cross-adsorbed and exhibits minimum reactivity to bovine, chicken, goat, human, mouse, pig and rat. For Western blot applications, 5% non-fat dry milk in PBST or TBST is recommended for blocking and incubation of antibodies. BSA is not recommended.
Specifications
Specifications
| Antigen | Rabbit IgG (H+L) Cross-Adsorbed |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | DyLight 488 |
| Formulation | PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4 |
| Host Species | Donkey |
| Immunogen | Rabbit IgG-heavy and light chain. |
| Purification Method | Antigen affinity chromatography |
| Quantity | 500 μg |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.