Learn More
Jackson Immuno Research Labs Biotin-SP (long spacer) AffiniPure Donkey Anti-Rat IgG (H+L), .5ml
SDP
Catalog No. NC9177929
Encompass_Preferred
Biotin-SP-AffiniPure Donkey Anti-Rat IgG (H+L) (min X Bov,Ck,Gt,GP,Sy Hms,Hrs,Hu,Ms,Rb,Shp Sr Prot)
Catalog No. NC9177929 Supplier Jackson Immuno Research Labs Supplier No. 712065153
Biotin-SP-AffiniPure Donkey Anti-Rat IgG (H+L) (min X Bov,Ck,Gt,GP,Sy Hms,Hrs,Hu,Ms,Rb,Shp Sr Prot)