Learn More
Jackson Immuno Research Labs Alexa Fluor 647 AffiniPure Donkey Anti-Rat IgG (H+L), .5mg
SDP
Catalog No. NC0692397
Encompass_Preferred
Alexa Fluor 647-AffiniPure Donkey Anti-Rat IgG (H+L) (min X Bov,Ck,Gt,GP,Sy Hms,Hrs,Hu,Ms,Rb,Shp Sr Prot)
Catalog No. NC0692397 Supplier Jackson Immuno Research Labs Supplier No. 712605153
Alexa Fluor 647-AffiniPure Donkey Anti-Rat IgG (H+L) (min X Bov,Ck,Gt,GP,Sy Hms,Hrs,Hu,Ms,Rb,Shp Sr Prot)