Learn More
Jackson Immuno Research Labs Cy™3 AffiniPure F(ab')₂ Fragment Donkey Anti-Rat IgG (H+L), .3mg
SDP
Catalog No. NC0810836
Encompass_Preferred
Cy3-AffiniPure F(ab')2 Fragment Donkey Anti-Rat IgG (H+L) (min X Bov,Ck,Gt,GP,Sy Hms,Hrs,Hu,Rb,Shp Sr Prot)
Catalog No. NC0810836 Supplier Jackson Immuno Research Labs Supplier No. 712166150
Cy3-AffiniPure F(ab')2 Fragment Donkey Anti-Rat IgG (H+L) (min X Bov,Ck,Gt,GP,Sy Hms,Hrs,Hu,Rb,Shp Sr Prot)